Learn More
Cayman Chemical Geldanamycin, Purity: ≥98%, CAS: 30562-34-6, 10 mg
SDP
Catalog No. 502955810
Encompass_Preferred
A benzoquinone ansamycin antibiotic which binds Hsp90 and its paralog GRP94; indirectly affects diverse cellular processes, including gene expression, cell proliferation, apoptosis, and angiogenesis; inhibits c-jun expression in HT29 cells (IC50 = 75 nM); shows promise in cancer therapy
Catalog No. 50-295-5810 Supplier Cayman Chemical Supplier No. 1335510 MG
A benzoquinone ansamycin antibiotic which binds Hsp90 and its paralog GRP94; indirectly affects diverse cellular processes, including gene expression, cell proliferation, apoptosis, and angiogenesis; inhibits c-jun expression in HT29 cells (IC50 = 75 nM); shows promise in cancer therapy