Learn More
Hach Company Gelex Secondary Turbidity Standard, 0-200 NTU
Catalog No. 501996279 Shop All Hach Company Products
Encompass
Gelex Secondary Turbidity Standard, 0-200 NTU, for 2100N, 2100N IS, 2100AN, 2100AN IS Turbidimeters.
Catalog No. 50-199-6279 Supplier Hach Company Supplier No. 2589103
Gelex Secondary Turbidity Standard, 0-200 NTU, for 2100N, 2100N IS, 2100AN, 2100AN IS Turbidimeters.