Learn More
Hach Company Gelex Secondary Turbidity Standard, 200-4000 NTU
Catalog No. 501996280 Shop All Hach Company Products
Encompass
Gelex Secondary Turbidity Standard, 200-4000 NTU, for 2100N, 2100AN, 2100AN IS Turbidimeter.
Catalog No. 50-199-6280 Supplier Hach Company Supplier No. 2589104
Gelex Secondary Turbidity Standard, 200-4000 NTU, for 2100N, 2100AN, 2100AN IS Turbidimeter.