Learn More
Hach Company Gelex Secondary Turbidity Standards Kit, 0 - 10000 NTU
Catalog No. 501996282 Shop All Hach Company Products
Encompass
Gelex Secondary Standards Kit, for 2100AN Laboratory Turbidimeter. Includes: 1 sealed vial each, Stray Light Standard, 0-2 NTU, 0-20 NTU, 0-200 NTU, 200-4000 NTU, 4000-10000 NTU. For 2100N, 2100N IS, 2100AN, 2100AN IS.
Catalog No. 50-199-6282 Supplier Hach Company Supplier No. 2589200
Gelex Secondary Standards Kit, for 2100AN Laboratory Turbidimeter. Includes: 1 sealed vial each, Stray Light Standard, 0-2 NTU, 0-20 NTU, 0-200 NTU, 200-4000 NTU, 4000-10000 NTU. For 2100N, 2100N IS, 2100AN, 2100AN IS.