Learn More
Hach Company Gelex Secondary Turbidity Standards Kit, 2100N IS
Catalog No. 501996378 Shop All Hach Company Products
Encompass
Gelex Secondary Turbidity Standard Kit includes 1 sealed vial each, Stray Light Standard, 0-2 NTU, 0-20 NTU, 0-200 NTU.
Catalog No. 50-199-6378 Supplier Hach Company Supplier No. 2620000
Gelex Secondary Turbidity Standard Kit includes 1 sealed vial each, Stray Light Standard, 0-2 NTU, 0-20 NTU, 0-200 NTU.