Learn More
Hach Company Gelex Secondary Standards Kit, 2100 Series Portable Turbidimeters
Catalog No. 501996048 Shop All Hach Company Products
Encompass
Gelex Secondary Standards Kit, for 2100P and 2100Q Series Portable Turbidimeters (includes 1 sealed vial each, 0-10 NTU, 10-100 NTU, 100-1000 NTU).
Catalog No. 50-199-6048 Supplier Hach Company Supplier No. 2464105
Gelex Secondary Standards Kit, for 2100P and 2100Q Series Portable Turbidimeters (includes 1 sealed vial each, 0-10 NTU, 10-100 NTU, 100-1000 NTU).