Learn More
Biotium GelGreen Agarose LE
Supplier:  Biotium 4103050G
Encompass_Preferred
GelGreen Agarose LE is our ultra-pure molecular biology grade LE agarose pre-coated with GelGreen Nucleic Acid Gel Stain. With GelGreen Agarose, there is no need to handle concentrated fluorescent dye while preparing your gel, for greater convenience and safety. GelGreen Agarose LE has low electroendosmosis (EEO) for high electrophoretic mobility. This agarose has excellent performance for analytical or preparative nucleic acid electrophoresis and blotting. It is suitable for preparing 0.8%-2% gels in TAE or TBE buffer. GelGreen Agarose also gives excellent results at percentages between 0.8% (0.8X GelGreen) up to 2% (2X GelGreen). GelGreen is a sensitive, stable, and environmentally safe green fluorescent nucleic acid dye specifically designed for gel staining. In a 1% GelGreen Agarose gel, the final GelGreen concentration is 1X.
Catalog No. 50-196-4680