Learn More
AMPLYUS LLC miniPCR bio GelGreen Agarose Tabs™, 8 tablets
SDP
Catalog No. 502237987
Encompass
All-in-one tabs with pre-measured agarose, GelGreen nucleic acid stain, and the TBE buffer needed to cast electrophoresis gels. GelGreen Agarose Tabs™ facilitate DNA or RNA visualization under safe blue light. 8 tablets.
This item is not returnable. View return policy
Catalog No. 50-223-7987 Supplier AMPLYUS LLC Supplier No. RG150011
All-in-one tabs with pre-measured agarose, GelGreen nucleic acid stain, and the TBE buffer needed to cast electrophoresis gels. GelGreen Agarose Tabs™ facilitate DNA or RNA visualization under safe blue light. 8 tablets.