Learn More
AMPLYUS LLC miniPCR bio GelGreen Nucleic Acid Gel Stain, 10,000X in Water, 200 µl
SDP
Catalog No. NC1555630
Encompass_Preferred
A sensitive, stable green fluorescent nucleic acid dye designed for precast or post-electrophoresis staining of dsDNA, ssDNA or RNA in agarose gels. GelGreen offers more sensitivity and stability (both hydrolytic and thermal) than SYBR dyes. GelGreen is compatible with gel purification, restriction digest, sequencing and cloning.
Catalog No. NC1555630 Supplier AMPLYUS LLC Supplier No. RG155001
A sensitive, stable green fluorescent nucleic acid dye designed for precast or post-electrophoresis staining of dsDNA, ssDNA or RNA in agarose gels. GelGreen offers more sensitivity and stability (both hydrolytic and thermal) than SYBR dyes. GelGreen is compatible with gel purification, restriction digest, sequencing and cloning.