Learn More
Sigma Aldrich Fine Chemicals Biosciences Gel Loading Buffer, 5 mL
Catalog No. NC0554379
Encompass
Gel loading buffer is used as a tracking dye during electrophoresis. The dye has a slight negative charge and will migrate the same direction as DNA, allowing the user to monitor the progress of molecules moving through the gel. The rate of migration varies with gel composition. Dilute 1:3 to 1:6 with sample before loading.
Catalog No. NC0554379 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. G25265ML
Gel loading buffer is used as a tracking dye during electrophoresis. The dye has a slight negative charge and will migrate the same direction as DNA, allowing the user to monitor the progress of molecules moving through the gel. The rate of migration varies with gel composition. Dilute 1:3 to 1:6 with sample before loading.