Learn More
Medchemexpress LLC Gellan gum | 71010-52-1 | MFCD00131909 | 97.0% | Polymer | 1G
SDP
Catalog No. 5000236116
Encompass_Preferred
Gellan gum is a linear microbial exopolysaccharide used as a gelling agent and scaffold for cell culture and tissue engineering. It forms firm, transparent gels in the presence of cations and offers biocompatibility, biodegradability, and low toxicity, making it suitable for soft tissue and load-bearing scaffold applications.
  • Provides biocompatible and biodegradable gel scaffolds for cell culture.
  • Forms stable, ion-responsive gels in the presence of cations.
  • Available as a white to off-white powder for easy handling and dissolution.
  • Manufacturer specification purity 97.0% for research use.
  • Suitable for tissue engineering, cell scaffolding, and agar replacement in culture.
  • Supplied in small research pack sizes for laboratory use.
Catalog No. 50-002-36116 Supplier Medchemexpress LLC Supplier No. HY1258701G
Gellan gum is a linear microbial exopolysaccharide used as a gelling agent and scaffold for cell culture and tissue engineering. It forms firm, transparent gels in the presence of cations and offers biocompatibility, biodegradability, and low toxicity, making it suitable for soft tissue and load-bearing scaffold applications.
  • Provides biocompatible and biodegradable gel scaffolds for cell culture.
  • Forms stable, ion-responsive gels in the presence of cations.
  • Available as a white to off-white powder for easy handling and dissolution.
  • Manufacturer specification purity 97.0% for research use.
  • Suitable for tissue engineering, cell scaffolding, and agar replacement in culture.
  • Supplied in small research pack sizes for laboratory use.