Learn More
Medchemexpress LLC Gellan gum | 71010-52-1 | MFCD00131909 | 5G
SDP
Catalog No. 5000235324
Encompass_Preferred
Gellan gum for cell culture is a linear microbial exopolysaccharide used as an agar substitute and as a scaffold material in cell and tissue culture. It forms transparent, stable gels at low concentrations in the presence of cations, and is valued for biocompatibility, biodegradability, and low toxicity.
  • Forms transparent gels at low concentrations in the presence of cations.
  • Biocompatible and biodegradable, with low toxicity.
  • Serves as an agar substitute for cell and plant tissue culture.
  • Powder form for straightforward dissolution and media preparation.
  • Batch COA reports gellan content 85.0-108.0 and gel strength ~1,408 g/cm2 (example batch).
Catalog No. 50-002-35324 Supplier Medchemexpress LLC Supplier No. HY1258705G
Gellan gum for cell culture is a linear microbial exopolysaccharide used as an agar substitute and as a scaffold material in cell and tissue culture. It forms transparent, stable gels at low concentrations in the presence of cations, and is valued for biocompatibility, biodegradability, and low toxicity.
  • Forms transparent gels at low concentrations in the presence of cations.
  • Biocompatible and biodegradable, with low toxicity.
  • Serves as an agar substitute for cell and plant tissue culture.
  • Powder form for straightforward dissolution and media preparation.
  • Batch COA reports gellan content 85.0-108.0 and gel strength ~1,408 g/cm2 (example batch).