Learn More
Medchemexpress LLC Gellan gum | 71010-52-1 | MFCD00131909 | 85.0-108.0% | Approximately 500,000 g/mol | No single chemical formula (polymeric polysaccharide) | 500MG
SDP
Catalog No. 5000235908
Encompass_Preferred
Gellan gum for cell culture is a microbial exopolysaccharide supplied as a white to off-white powder. It acts as an agar substitute and scaffold material for tissue engineering and cell culture, forming gels with tunable mechanical properties. The supplied material is characterized by a reported gellan content of 85.0-108.0 and a pH range of 5-8 per the certificate of analysis.
  • Microbial exopolysaccharide suitable as an agar substitute.
  • Forms gels with tunable mechanical properties for scaffolding.
  • White to off-white powder for easy handling and dissolution.
  • Appropriate for soft tissue and load-bearing constructs.
  • Certificate of analysis documents purity and pH specifications.
  • Small research-scale pack suitable for laboratory use.
Catalog No. 50-002-35908 Supplier Medchemexpress LLC Supplier No. HY125870500MG
Gellan gum for cell culture is a microbial exopolysaccharide supplied as a white to off-white powder. It acts as an agar substitute and scaffold material for tissue engineering and cell culture, forming gels with tunable mechanical properties. The supplied material is characterized by a reported gellan content of 85.0-108.0 and a pH range of 5-8 per the certificate of analysis.
  • Microbial exopolysaccharide suitable as an agar substitute.
  • Forms gels with tunable mechanical properties for scaffolding.
  • White to off-white powder for easy handling and dissolution.
  • Appropriate for soft tissue and load-bearing constructs.
  • Certificate of analysis documents purity and pH specifications.
  • Small research-scale pack suitable for laboratory use.