Learn More
Bioworld Gellan Gum Powder, 250 g
Catalog No. 50254744
Encompass_Preferred
Gellan Gum PowderSynonym: Culturegel™ Type I: Biotech GradePlant tissue culture testedGel Strength: 800 g/cm2
Catalog No. 50-254-744 Supplier Bioworld Supplier No. 306300901
Gellan Gum PowderSynonym: Culturegel™ Type I: Biotech GradePlant tissue culture testedGel Strength: 800 g/cm2