Learn More
Bioworld Gellan Gum Powder, 1 kg
Catalog No. 50254746
Encompass_Preferred
Gellan Gum PowderSynonym: Culturegel™ Type I: Biotech GradePlant tissue culture testedGel Strength: 800 g/cm2
Catalog No. 50-254-746 Supplier Bioworld Supplier No. 306300903
Gellan Gum PowderSynonym: Culturegel™ Type I: Biotech GradePlant tissue culture testedGel Strength: 800 g/cm2