Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gellan Gum Powder, MP Biomedicals™

Catalog No. MP021801065 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 g
500 g
1 kg
5 kg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP021801065 500 g
MP021801061 100 g
MP218010601 1 kg
MP218010605 5 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. MP021801065 Supplier MP Biomedicals, Inc Supplier No. 02180106.5
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Used as a gelling agent in place of agar in microbiological media. Gellan Gum Powder, MP Biomedicals is a water-soluble polysaccharide produced by Pseudomonas elodea, a bacterium. It is used in a variety of immobilization matricies and as a food additive,

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 71010-52-1
Molecular Formula (C24H38O20)A
Synonym Agar substitute gelling agent

Specifications

Quantity 500 g
Physical Form Powder
Recommended Storage Store at Room Temperature (15–30°C).
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.