Learn More
Bioworld Gellan Gum Powder, 500 g
Catalog No. 50254745
Encompass_Preferred
Gellan Gum PowderSynonym: Culturegel™ Type I: Biotech GradePlant tissue culture testedGel Strength: 800 g/cm2
Catalog No. 50-254-745 Supplier Bioworld Supplier No. 306300902
Gellan Gum PowderSynonym: Culturegel™ Type I: Biotech GradePlant tissue culture testedGel Strength: 800 g/cm2