Learn More
PLANT CELL TECHNOLOGY Gelling Agent, Plant Tissue Culture, Supreme TC Grade Agar, Off-White to Tan Powder, 1kg Pack
Catalog No. NC3905191
Encompass_Preferred
Supreme TC Grade Agar is a high-purity gelling agent for plant tissue culture. This off-white to tan powder dissolves easily with heat to form an opaque gel, supporting versatile plant tissue culture applications. Supplied in 1kg units, this agar is ideal for research labs, commercial greenhouses, and educational applications. Shelf stable under recommended conditions, ensuring consistent gel strength and reliable culture support.
Catalog No. NC3905191 Supplier PLANT CELL TECHNOLOGY Supplier No. CNA0601KG
Supreme TC Grade Agar is a high-purity gelling agent for plant tissue culture. This off-white to tan powder dissolves easily with heat to form an opaque gel, supporting versatile plant tissue culture applications. Supplied in 1kg units, this agar is ideal for research labs, commercial greenhouses, and educational applications. Shelf stable under recommended conditions, ensuring consistent gel strength and reliable culture support.