Learn More
Cole-Parmer Cole-Parmer Gel-Loading Tips, 1 to 200 µL, 5 Racks of 204 Tips; 1020 Tips/pk
Catalog No. NC1888283 Shop All Cole-Parmer Products
Encompass_Preferred
These polypropylene tips are rounded to fit easily into small instrument openings. Use these microcapillary tips for DNA sequencing, loading SDS PAGE gels, and other critical sampling applications.
Catalog No. NC1888283 Supplier Cole-Parmer Supplier No. 2571312
These polypropylene tips are rounded to fit easily into small instrument openings. Use these microcapillary tips for DNA sequencing, loading SDS PAGE gels, and other critical sampling applications.