Learn More
Mitegen Gel Loading Pipette Tips, For Precise Sample Loading & Liquid Transfers, 200 Tips per Pack
SDP
Catalog No. 501939584
Encompass
These Gel Loading Pipette Tips are engineered for exceptional precision and accuracy in various laboratory applications, particularly for gel electrophoresis and small volume liquid transfers. Designed with a fine, extended tip, they facilitate easy and controlled access into narrow wells, ensuring precise sample loading without cross-contamination or damage to the gel matrix. Manufactured to high quality standards, these tips offer reliable and reproducible performance for sensitive assays. Their design makes them ideal for tasks such as loading DNA or protein samples into gels, performing micro-scale mixing, or transferring minute liquid aliquots in crystallography, molecular biology, and biochemistry research. Providing a smooth and consistent flow, these tips contribute to enhanced experimental efficiency and reliable results. Each pack contains 200 tips, offering a convenient supply for demanding laboratory workflows.
Catalog No. 50-193-9584 Supplier Mitegen Supplier No. GLPT1
These Gel Loading Pipette Tips are engineered for exceptional precision and accuracy in various laboratory applications, particularly for gel electrophoresis and small volume liquid transfers. Designed with a fine, extended tip, they facilitate easy and controlled access into narrow wells, ensuring precise sample loading without cross-contamination or damage to the gel matrix. Manufactured to high quality standards, these tips offer reliable and reproducible performance for sensitive assays. Their design makes them ideal for tasks such as loading DNA or protein samples into gels, performing micro-scale mixing, or transferring minute liquid aliquots in crystallography, molecular biology, and biochemistry research. Providing a smooth and consistent flow, these tips contribute to enhanced experimental efficiency and reliable results. Each pack contains 200 tips, offering a convenient supply for demanding laboratory workflows.