Learn More
Advanced Biomatrix Preformulated GelMA methacrylated gelatin 20% concentration
Catalog No. 502255981
Encompass_Preferred
GelMA is porcine gelatin functionalized with methacrylate groups. GelMA 20% is a concentrated solution at physiological pH and osmolarity, to be used as a base in bioink formulation. This material has a ~50% degree of methacrylation.
Catalog No. 50-225-5981 Supplier Advanced Biomatrix Supplier No. IKG125000005
GelMA is porcine gelatin functionalized with methacrylate groups. GelMA 20% is a concentrated solution at physiological pH and osmolarity, to be used as a base in bioink formulation. This material has a ~50% degree of methacrylation.