Learn More
Advanced Biomatrix GelMA A Methacrylated Gelatin 50% degree of methacrylation and Alginate bioink with LAP for extrusion printers 3 x 3 mL
Supplier:  Advanced Biomatrix IK3521020303
Encompass_Preferred
GelMA A is a blend of GelMA and alginate, offering a larger printability window compared to pure GelMA bioinks due to the softening provided by the alginate. GelMA A can be stabilized through photocrosslinking, or through the addition of the ionic crosslinking solution.
Catalog No. 50-225-5959