Learn More
Advanced Biomatrix GelMA Methacrylated Gelatin 50% degree of methacrylation bioink with LAP for extrusion printers 3 x 3 mL
Catalog No. 502255957
Encompass_Preferred
Gelatin methacrylate (GelMA) is a gelatin-based bioink that provides mammalian cells with the essential properties of their native environment. GelMA is modified with methacryloyl groups to crosslink with exposure to a 405 nm photocuring system. GelMA bioink comes pre-mixed with a 0.25 percent LAP photoinitiator.
This item is not returnable. View return policy
Catalog No. 50-225-5957 Supplier Advanced Biomatrix Supplier No. IK3051020303
Gelatin methacrylate (GelMA) is a gelatin-based bioink that provides mammalian cells with the essential properties of their native environment. GelMA is modified with methacryloyl groups to crosslink with exposure to a 405 nm photocuring system. GelMA bioink comes pre-mixed with a 0.25 percent LAP photoinitiator.