Learn More
Advanced Biomatrix GelMA C Methacrylated Gelatin 50% degree of methacrylation and Cellulose bioink with LAP for extrusion printers 3 x 3 mL
Supplier:  Advanced Biomatrix IKG5L3000303
Encompass_Preferred
GelMA C combines GelMA with nanofibrillated cellulose. This unique blend offers smooth printability at ambient conditions and low extrusion pressures using a wide range of nozzle diameters, while retaining universal cell attachment motifs from GelMA. GelMA C crosslinks quickly through photocuring after printing.
Catalog No. 50-225-5961