Learn More
Med Vet International Miltex Gelpi Retractor, 6.75in., Grip Lock Ratchet
Catalog No. 502831136
Encompass_Preferred
The Miltex 11-366 Gelpi retractor is made of OR grade German stainless steel and measures 6.75" in length. The retractor has a grip lock ratchet.
Catalog No. 50-283-1136 Supplier Med Vet International Supplier No. MIL11366
The Miltex 11-366 Gelpi retractor is made of OR grade German stainless steel and measures 6.75" in length. The retractor has a grip lock ratchet.