Learn More
Med Vet International Miltex Vantage Gelpi Retractor, 6.75in., Grip Lock Ratchet
Catalog No. 502830418
Encompass_Preferred
The Miltex Vantage V911-366 Gelpi retractor is made from high quality Japanese stainless steel and measures 6.75" in length. The retractor has a grip lock ratchet.
Catalog No. 50-283-0418 Supplier Med Vet International Supplier No. MILV911366
The Miltex Vantage V911-366 Gelpi retractor is made from high quality Japanese stainless steel and measures 6.75" in length. The retractor has a grip lock ratchet.