Learn More
Research Products International Corp Gelplate-Clean Spray, 2 x 250 ml
SDP
Catalog No. 50212677
Encompass
GELPLATE-Clean is specifically formulated for cleaning electrophoresis gel plates. Removes proteins, nucleic acids, fats, lipids, radioisotopes and other contaminants from electrophoresis gel plates. Suitable for sequencing, protein and mini electrophoresis gel plates. Easy to use, spray and wipe gel plate. Regular use of GELPLATE-Clean improves quality of electrophoresis results-prevents poor electrophoresis migration, band distortion and poor image development.
Supplied as two 250 ml spray bottles.
Catalog No. 50-212-677 Supplier Research Products International Corp Supplier No. 249865
GELPLATE-Clean is specifically formulated for cleaning electrophoresis gel plates. Removes proteins, nucleic acids, fats, lipids, radioisotopes and other contaminants from electrophoresis gel plates. Suitable for sequencing, protein and mini electrophoresis gel plates. Easy to use, spray and wipe gel plate. Regular use of GELPLATE-Clean improves quality of electrophoresis results-prevents poor electrophoresis migration, band distortion and poor image development.
Supplied as two 250 ml spray bottles.