Learn More
Biotium GelRed Agarose LE
Supplier:  Biotium 4102950G
Encompass_Preferred
GelRed Agarose LE is our ultra-pure molecular biology grade LE agarose pre-coated with GelRed Nucleic Acid Gel Stain. With GelRed Agarose, there is no need to handle concentrated fluorescent dye while preparing your gel, for greater convenience and safety. GelRed Agarose LE has low electroendosmosis (EEO) for high electrophoretic mobility. This agarose demonstrates excellent performance for analytical or preparative nucleic acid electrophoresis and blotting. It is suitable for preparing 0.8%-2% gels in TAE or TBE buffer. In a 1% GelRed Agarose gel, the final GelRed concentration is 1X. GelRed Agarose also gives excellent results at percentages between 0.8% (0.8X GelRed) up to 2% (2X GelRed). GelRed and EtBr have virtually the same spectra, so you can directly replace EtBr with GelRed without changing your existing imaging system. Gel staining with GelRed is compatible with downstream applications such as sequencing and cloning.
Catalog No. 50-196-4678