Learn More
Biotium PAGE GelRed Nucleic Acid Gel Stain 0.1 mL
Catalog No. NC1712531
Encompass_Preferred
PAGE GelRed is a non-toxic, non-mutagenic red DNA gel stain specifically formulated to stain DNA in polyacrylamide gels. PAGE GelRed can be imaged using a 254 nm UV transilluminator with an ethidium bromide filter.
Catalog No. NC1712531 Supplier Biotium Supplier No. 41008T
PAGE GelRed is a non-toxic, non-mutagenic red DNA gel stain specifically formulated to stain DNA in polyacrylamide gels. PAGE GelRed can be imaged using a 254 nm UV transilluminator with an ethidium bromide filter.