Learn More
Gold Biotechnology Inc GelRed Nucleic Acid Gel Stain, 10,000X in Water 500 µL
SDP
Catalog No. NC2943351
Encompass_Preferred
GelRed is an ultra-sensitive, extremely stable and environmentally safe fluorescent nucleic acid dye designed to replace the highly toxic ethidium bromide (EtBr) for staining dsDNA, ssDNA or RNA in agarose gels or polyacrylamide gels. GelRed is far more sensitive than EtBr without requiring a destaining step. GelRed and EtBr have virtually the same spectra, so you can directly replace EtBr with GelRed without changing your existing imaging system.
Catalog No. NC2943351 Supplier Gold Biotechnology Inc Supplier No. G725500
GelRed is an ultra-sensitive, extremely stable and environmentally safe fluorescent nucleic acid dye designed to replace the highly toxic ethidium bromide (EtBr) for staining dsDNA, ssDNA or RNA in agarose gels or polyacrylamide gels. GelRed is far more sensitive than EtBr without requiring a destaining step. GelRed and EtBr have virtually the same spectra, so you can directly replace EtBr with GelRed without changing your existing imaging system.