Learn More
Biotium GelRed Prestain Plus 6X DNA Loading Dye, 1mL
Catalog No. NC1712532
Encompass_Preferred
6X DNA loading buffer that includes ultra-sensitive, non-toxic GelRed dye. This is an improved version of our original GelRed prestain loading buffer, formulated to minimize DNA migration shift.
Catalog No. NC1712532 Supplier Biotium Supplier No. 41011
6X DNA loading buffer that includes ultra-sensitive, non-toxic GelRed dye. This is an improved version of our original GelRed prestain loading buffer, formulated to minimize DNA migration shift.