Learn More
Sigma Aldrich Fine Chemicals Biosciences GelRedR Nucleic Acid Stain
Catalog No. 502900318
Encompass
GelRed is a sensitive stable and environmentally safe fluorescent nucleic acid dye designed to replace the highly toxic ethidium bromide (EtBr) for staining dsDNA ssDNA or RNA in agarose gels or polyacrylamide gels. GelRed and EtBr have virtually the same spectra (figure 1) so you can directly replace EtBr with GelRed without changing your existing imaging system. In addition GelRed is far more sensitive than EtBr (figure 2).The dye is noncytotoxic and non-mutagenic at concentrations well above the working concentrations used in gel staining. GelRed successfully passed environmental safety tests in compliance with CCR Title 22 Hazardous Waste Characterization under which GelRed is not classified as hazardous waste.Spectral PropertiesAbsorbance Standard Transilluminator (302 or 312 nm)Emission Ethidium Bromide SYBR or GelStar filter
Catalog No. 50-290-0318 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. SCT122
GelRed is a sensitive stable and environmentally safe fluorescent nucleic acid dye designed to replace the highly toxic ethidium bromide (EtBr) for staining dsDNA ssDNA or RNA in agarose gels or polyacrylamide gels. GelRed and EtBr have virtually the same spectra (figure 1) so you can directly replace EtBr with GelRed without changing your existing imaging system. In addition GelRed is far more sensitive than EtBr (figure 2).The dye is noncytotoxic and non-mutagenic at concentrations well above the working concentrations used in gel staining. GelRed successfully passed environmental safety tests in compliance with CCR Title 22 Hazardous Waste Characterization under which GelRed is not classified as hazardous waste.Spectral PropertiesAbsorbance Standard Transilluminator (302 or 312 nm)Emission Ethidium Bromide SYBR or GelStar filter