Learn More
Gold Biotechnology Inc GelRedTM Nucl Acid Gel Stain
SDP
Catalog No. 5000562246
Encompass_Preferred
GelRed is an ultra-sensitive extremely stable and environmentally safe fluorescent nucleic acid dye designed to replace the highly toxic ethidium bromide (EtBr) for staining dsDNA ssDNA or RNA in agarose gels or polyacrylamide gels GelRed is far more sensitive than EtBr without requiring a destaining step GelRed and EtBr have virtually the same spectra so you can directly replace EtBr with GelRed without changing your existing imaging system
Catalog No. 50-005-62246 Supplier Gold Biotechnology Inc Supplier No. G7252
GelRed is an ultra-sensitive extremely stable and environmentally safe fluorescent nucleic acid dye designed to replace the highly toxic ethidium bromide (EtBr) for staining dsDNA ssDNA or RNA in agarose gels or polyacrylamide gels GelRed is far more sensitive than EtBr without requiring a destaining step GelRed and EtBr have virtually the same spectra so you can directly replace EtBr with GelRed without changing your existing imaging system