Learn More
Research Products International Corp Gelrite, 5 Kilograms
SDP
Catalog No. 50488684
Encompass_Preferred
Naturally derived gelling polymer that can be used in place of agar. Gelrite forms clear, rigid gels at approximately half the use level of agar in presence of soluble salts like Mg2+ and Ca2+.
Catalog No. 50-488-684 Supplier Research Products International Corp Supplier No. G350205000.0
Naturally derived gelling polymer that can be used in place of agar. Gelrite forms clear, rigid gels at approximately half the use level of agar in presence of soluble salts like Mg2+ and Ca2+.