Learn More
Crescent Chemical Co Inc GELRITE
SDP
Catalog No. 50247620
Encompass_Preferred
Gelrite Polysaccharide produced by bacteria. Useful alternative to agar for the in vitro culture of many plants. Yields very clear gels. Constant quality from lot to lot. Size - 1KG Storage Conditions - +15 C TO +30 C Catalog Number - 22
Catalog No. 50-247-620 Supplier Crescent Chemical Co Inc Supplier No. SE22168.02
Gelrite Polysaccharide produced by bacteria. Useful alternative to agar for the in vitro culture of many plants. Yields very clear gels. Constant quality from lot to lot. Size - 1KG Storage Conditions - +15 C TO +30 C Catalog Number - 22