Learn More
Crescent Chemical Co Inc GELRITE
SDP
Catalog No. 50247619
Encompass_Preferred
Gelrite Polysaccharide produced by bacteria. Useful alternative to agar for the in vitro culture of many plants. Yields very clear gels. Constant quality from lot to lot. Size - 250G Storage Conditions - +15 C TO +30 C Catalog Number - 2
Catalog No. 50-247-619 Supplier Crescent Chemical Co Inc Supplier No. SE22168.01
Gelrite Polysaccharide produced by bacteria. Useful alternative to agar for the in vitro culture of many plants. Yields very clear gels. Constant quality from lot to lot. Size - 250G Storage Conditions - +15 C TO +30 C Catalog Number - 2