Learn More
Research Products International Corp Gelrite, 500 Grams
SDP
Supplier:  Research Products International Corp G35020500.0
Encompass_Preferred
Naturally derived gelling polymer that can be used in place of agar. Gelrite forms clear, rigid gels at approximately half the use level of agar in presence of soluble salts like Mg2+ and Ca2+.
Catalog No. 50-213-334