Learn More
Medchemexpress LLC Gelsevirine | 38990-03-3 | 5 MG
SDP
Catalog No. 5000420079
Encompass_Preferred
Gelsevirine is the major alkaloid in Gelsemium elegans with potent anxiolytic effects. The anxiolytic mechanism of Gelsevirine may involve the agonist action of the glycine receptor in the brain. Gelsevirine also exhibits anti-proliferation activity with IC50 values of 1.41 mM and 1.22 mM for SW480 cells and MGC80-3 cells, respectively.
  • Major alkaloid in Gelsemium elegans
  • Potent anxiolytic effects
  • Anxiolytic mechanism may involve agonist action of glycine receptor
  • Exhibits anti-proliferation activity
Catalog No. 50-004-20079 Supplier Medchemexpress LLC Supplier No. HYN39405MG
Gelsevirine is the major alkaloid in Gelsemium elegans with potent anxiolytic effects. The anxiolytic mechanism of Gelsevirine may involve the agonist action of the glycine receptor in the brain. Gelsevirine also exhibits anti-proliferation activity with IC50 values of 1.41 mM and 1.22 mM for SW480 cells and MGC80-3 cells, respectively.
  • Major alkaloid in Gelsemium elegans
  • Potent anxiolytic effects
  • Anxiolytic mechanism may involve agonist action of glycine receptor
  • Exhibits anti-proliferation activity