Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ Geltrex™ Flex LDEV-Free Organoid-Qualified Reduced Growth Factor Basement Membrane Matrix

Catalog No. A4000046902
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mL
5 mL
10 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A4000046902 5 mL
A4000046901 1 mL
A4000046903 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. A4000046902 Supplier Gibco™ Supplier No. A4000046902
Add to Cart
Edge
Add to Cart

Geltrex Flex LDEV-Free Organoid-Qualified Reduced Growth Factor Basement Membrane Matrix is a soluble basement membrane matrix purified from EHS tumor cells used for the culture of tumoroids and organoids. Each lot is tested for the culture of mouse intestinal, liver ductal, airway, and mouse small intestinal organoids.

Gibco Geltrex Flex LDEV-Free Organoid-Qualified Reduced Growth Factor Basement Membrane Matrix is a soluble form of basement membrane that is purified from Engelbreth-Holm-Swarm (EHS) tumor cells. The major components of Geltrex Flex matrix include laminin, collagen IV, entactin, and heparan sulfate proteoglycan.

Recommended for culturing tumoroids and other organoids, organoid-qualified Geltrex Flex matrix was used to grow human lung cancer tumoroids and human colorectal cancer tumoroids in suspension and embedded culture using the OncoPro Tumoroid Culture Medium Kit. For a detailed description of OncoPro tumoroid culture, see the OncoPro Tumoroid Culture Medium Kit User Guide (Pub. No. MAN0029372). To help ensure support of organoid culture, every lot of organoid-qualified Geltrex Flex matrix is quality control–tested for stable dome formation and growth of multiple mouse organoids.

Features of Geltrex Flex LDEV-Free Organoid-Qualified Reduced Growth Factor Basement Membrane Matrix

  • Lots tested to help ensure growth of mouse intestinal, liver, ductal, and airway organoids and the expansion of mouse small intestinal organoids for more than five passages with typical organoid budding morphology
  • Each lot tested for the absence of lactose dehydrogenase elevating virus (LDEV), mycoplasma, and a broad panel of viruses and microorganisms
  • Lots tested for endotoxin, maintenance of gel stability, and HUVEC tube formation
  • Growth factor–reduced to improve lot-to-lot consistency
  • Formulated without phenol red to minimize potential for estrogen-like effects
  • Provided in recyclable glass vials
  • 1-mL, 5-mL, 10-mL sizes for maximum flexibility. Use only what you need. Reduce need for extensive aliquoting and unwanted freeze-thaws.

For the culture of primary cells, such as epithelial cells, endothelial cells, smooth muscle cells, and angiogenesis assays, neurite outgrowth assays, or bile canaliculi formation assays with polarized hepatocytes, standard Geltrex Flex matrix can be used.

For culturing hESCs or iPSCs, it is recommended to use Geltrex Flex LDEV-Free hESC-Qualified Reduced Growth Factor Basement Membrane Matrix. Each lot of hESC-qualified Geltrex Flex matrix is tested with iPSCs and H9 hESCs for at least five passages to help ensure reliability and consistency in hESC/iPSC studies.

TRUSTED_SUSTAINABILITY

Specifications

Cell Type Human colorectal tumoroids, human lung tumoroids, mouse intestinal organoids, mouse liver ductal organoids, mouse airway organoids, HUVEC, primary epithelial cells, endothelial cells, smooth muscle cells, hepatocytes.
Classification Reduced Growth Factor
Concentration 9 to 13 mg/mL
Form Frozen
Product Type Basement Membrane Matrix
Serum Level Serum-free
Sterility Sterile
Without Additives No Phenol Red
Quantity 5 mL
Shipping Condition Dry Ice
Endotoxin Level ≤ 4.5 EU/mL
Purity or Quality Grade Cell Culture Grade
Species Mouse
Culture Type Adherent Cell Culture, Mammalian Cell Culture, 3D Cell Culture
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.