Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ Geltrex™ LDEV-Free Reduced Growth Factor Basement Membrane Matrix

Catalog No. p-9713000
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mL
5 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A1413202 5 mL
A1413201 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A1413202 Supplier Gibco™ Supplier No. A1413202
Only null left
Add to Cart
Edge
Add to Cart

A soluble form of basement membrane extracted from murine Engelbreth-Holm-Swarm (EHS) tumors. It is free of viruses, including lactose dehydrogenase elevating virus (LDEV), sometimes found in this type of preparation.

  • The major components of Geltrex™ matrix include laminin, collagen IV, entactin, and heparin sulfate proteoglycans
  • It is ideal for the promotion and maintenance of differentiated phenotypes in a variety of cell cultures including primary epithelial cells, endothelial cells, smooth muscle cells, and human induced pluripotent stem cells (iPSC)
  • Geltrex™ matrix offers, on average, a 2-fold reduction in several growth factors
  • As a result, analysis of specific growth factor effects in applications requiring basement membrane extract can be carried out in a controlled and reproducible format
  • Minimal protein concentration variation from lot-to-lot (∼15 mg/mL)
TRUSTED_SUSTAINABILITY

Specifications

Cell Type Variety of cultured cells, including primary epithelial cells, endothelial cells, smooth muscle cells, and human induced pluripotent stem cells (iPSC)
Classification Reduced Growth Factor
Concentration 12 to 18 mg/mL
Form Frozen
Product Type Basement Membrane Matrix
Serum Level Serum-free
Sterility Sterile
Without Additives No Phenol Red
Product Line Gibco
Quantity 5 mL
Shipping Condition Dry Ice
Endotoxin Level Low
Format Tube(s)
Purity or Quality Grade Cell Culture Grade
Species Mouse
Content And Storage Store at -20°C.
Culture Type Adherent Cell Culture
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.