Learn More
Medchemexpress LLC HY-Y1892 25g , Gelucire 44/14 CAS:121548-04-7 Purity:>98%
SDP
Catalog No. 502262003
Encompass_Preferred
HY-Y1892 25g Gelucire 44/14 CAS: 121548-04-7 Gelucire 44/14 is a potential and safe absorption enhancer for improving the absorption of poorly absorbable drugs including insulin and calcitonin by pulmonary delivery. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
This item is not returnable. View return policy
Catalog No. 50-226-2003 Supplier Medchemexpress LLC Supplier No. HYY189225G
HY-Y1892 25g Gelucire 44/14 CAS: 121548-04-7 Gelucire 44/14 is a potential and safe absorption enhancer for improving the absorption of poorly absorbable drugs including insulin and calcitonin by pulmonary delivery. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.