Learn More
Bioworld Gelzan™, 71010-52-1, 250g
Catalog No. NC9669831
Encompass_Preferred
Agar substitute produced from a bacterial substrate composed of glucaronic acid, rhamnose and glucose. Aids in the detection of microbial contaminants, Gel Strength: 400–700 g/cm2, Recommended concentration: 3.0–5.0 g/L, Gelling Temperature: 29–31°C, Loss on drying: 6–16%. Adjust Gelzan to the medium by means of a sieve to avoid clumping. Gelzan™ is a trademark of the Monsanto Company. Molecular Weight : 1,000 kDA. Solubility : Soluble in water, Appearance Form : Powder, ppearance Color : White to off-white. 250g
Catalog No. NC9669831 Supplier Bioworld Supplier No. 306340031
Agar substitute produced from a bacterial substrate composed of glucaronic acid, rhamnose and glucose. Aids in the detection of microbial contaminants, Gel Strength: 400–700 g/cm2, Recommended concentration: 3.0–5.0 g/L, Gelling Temperature: 29–31°C, Loss on drying: 6–16%. Adjust Gelzan to the medium by means of a sieve to avoid clumping. Gelzan™ is a trademark of the Monsanto Company. Molecular Weight : 1,000 kDA. Solubility : Soluble in water, Appearance Form : Powder, ppearance Color : White to off-white. 250g