Learn More
Electron Microscopy Sciences GEM 3 Facet Aluminum Back SE Safety Dispenser
SDP
Supplier:  Electron Microscopy Sciences 71967CS
Encompass_Preferred
Gem Scientific 3 facet blade have a sharp edge and better handling. Stainless Steel, .009" thick, 2 1/4" x 1/2" size, packed 100 blades in a Dispenser. These uncoated blades offer precision, sharpness and safety and come in a dispenser which offers you safety.,
Catalog No. 50-192-7058