Learn More
Aviva Systems Biology Corp GEM Antibody - C-terminal region (ARP42226_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP42226_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against GEM. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the C terminal region of human GEM. Alternate names: KIR, MGC26294ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-58150