Learn More
Structure Probe Inc GEM BLADE SINGLE EDGE 100PK
SDP
Catalog No. NC9009019
Encompass
GEM Scientific Blade, 40mm L x 20mm W x 0.23mm T along the cutting edge; up to 1mm along the top of the blade, Handy packs of ten are secured within an all plastic safety dispenser, 10/PK
Catalog No. NC9009019 Supplier Structure Probe Inc Supplier No. 05025MB
GEM Scientific Blade, 40mm L x 20mm W x 0.23mm T along the cutting edge; up to 1mm along the top of the blade, Handy packs of ten are secured within an all plastic safety dispenser, 10/PK