Learn More
Electron Microscopy Sciences Gem Runner™ Pop-Up Dispenser
SDP
Catalog No. 502090998
Encompass_Preferred
Gem Runner™ pop-up blade dispenser serves up single-edge blades at the touch of a button. It's compact, reliable, and reloadable. It speeds production while minimizing the risk of cuts. It's easily reloadable with a cartridge of single-edge blades: the empty cartridge slides out, and the full cartridge slides in. GEM2 and GEM3 can be used and are available in clean room ready packaging. When loaded with a cartridge of washed blades there is less opportunity for foreign material transfer or contamination from the blade to your finished product. Works with all GEM2 and GEM3 single-edge blades. *Blades sold separately.
Catalog No. 50-209-0998 Supplier Electron Microscopy Sciences Supplier No. 7198510
Gem Runner™ pop-up blade dispenser serves up single-edge blades at the touch of a button. It's compact, reliable, and reloadable. It speeds production while minimizing the risk of cuts. It's easily reloadable with a cartridge of single-edge blades: the empty cartridge slides out, and the full cartridge slides in. GEM2 and GEM3 can be used and are available in clean room ready packaging. When loaded with a cartridge of washed blades there is less opportunity for foreign material transfer or contamination from the blade to your finished product. Works with all GEM2 and GEM3 single-edge blades. *Blades sold separately.