Learn More
Electron Microscopy Sciences GEM Single Edge in Dispenser with Used Blade Storage
SDP
Catalog No. 5034099
Encompass_Preferred
These GEM single edge carbon steel razor blades (0.009" thick) are in a compact, safe, pop-up blade dispenser with an used blade storage. Safe, easy to remove blade when needed. Secure used blade storage in rear of unit. Clear see through cover reveals remaining blades. Push button dispenses blades. 100 blades in dispenser.
Catalog No. 50-340-99 Supplier Electron Microscopy Sciences Supplier No. 7195501
These GEM single edge carbon steel razor blades (0.009" thick) are in a compact, safe, pop-up blade dispenser with an used blade storage. Safe, easy to remove blade when needed. Secure used blade storage in rear of unit. Clear see through cover reveals remaining blades. Push button dispenses blades. 100 blades in dispenser.