Learn More
Medchemexpress LLC HY-109567 10mg Medchemexpress, Gemcabene CAS:183293-82-5 Purity:>98%
SDP
Catalog No. 501932343
Encompass_Preferred
Medchemexpress, HY-109567 10mg Gemcabene CAS:183293-82-5 Gemcabene (PD-72953), a first-in-class lipid-lowering agent, lowers low-density lipoprotein cholesterol (LDL-C), decreases triglycerides, and raises high-density lipoprotein cholesterol (HDL-C) and lowers pro-inflammatory acute-phase protein, C-reactive protein (CRP), exerting anti-inflammatory activity [1] [2] [3] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2343 Supplier Medchemexpress LLC Supplier No. HY10956710MG
Medchemexpress, HY-109567 10mg Gemcabene CAS:183293-82-5 Gemcabene (PD-72953), a first-in-class lipid-lowering agent, lowers low-density lipoprotein cholesterol (LDL-C), decreases triglycerides, and raises high-density lipoprotein cholesterol (HDL-C) and lowers pro-inflammatory acute-phase protein, C-reactive protein (CRP), exerting anti-inflammatory activity [1] [2] [3] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.