Learn More
Medchemexpress LLC HY-109567A 5mg Medchemexpress, Gemcabene (calcium) CAS:209789-08-2 Purity:>98%
SDP
Catalog No. 501932344
Encompass_Preferred
Medchemexpress, HY-109567A 5mg Gemcabene (calcium) CAS:209789-08-2 Gemcabene calcium (PD-72953 calcium), a first-in-class lipid-lowering agent, lowers low-density lipoprotein cholesterol (LDL-C), decreases triglycerides, and raises high-density lipoprotein cholesterol (HDL-C) and lowers pro-inflammatory acute-phase protein, C-reactive protein (CRP), exerting anti-inflammatory activity [1] [2] [3] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2344 Supplier Medchemexpress LLC Supplier No. HY109567A5MG
Medchemexpress, HY-109567A 5mg Gemcabene (calcium) CAS:209789-08-2 Gemcabene calcium (PD-72953 calcium), a first-in-class lipid-lowering agent, lowers low-density lipoprotein cholesterol (LDL-C), decreases triglycerides, and raises high-density lipoprotein cholesterol (HDL-C) and lowers pro-inflammatory acute-phase protein, C-reactive protein (CRP), exerting anti-inflammatory activity [1] [2] [3] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.