Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GeminiBio GemCell™ ANZ Origin Fetal Bovine Serum
SDP

Catalog No. 11852140 Shop All Gemini Bio Products
Change view
Click to view available options
Country of Origin:
Australia
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Country of Origin
11-852-140 Australia
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-852-140 Supplier GeminiBio Supplier No. 100700500ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Fully traceable serum from BSE/TSE-negligible regions for research cell culture and biomanufacturing.

  • Sourced exclusively from Australia and/or New Zealand, both BSE/TSE-negligible regions with minimal incidence of bovine infectious diseases including FMD and rabies
  • Collected at government-inspected facilities from healthy cattle born and raised in Australia and/or New Zealand that passed ante- and post-mortem veterinary inspection
  • Negligible risk of intra-herd mixing with animals from other countries due to geographic isolation
  • Meets stringent FBS requirements set by regulatory bodies worldwide
  • Triple 0.1 μm sterile-filtered for optimal purity
  • Mycoplasma-tested and virus-screened
  • Up to 5-year shelf life from date of manufacture
  • Ships on dry ice; store frozen at ≤ -10°C
  • Ideal for general cell culture, stem cell research, immunology, cancer research, biotechnology, and biomanufacturing
  • Manufactured under an ISO 13485:2016 certified quality system
  • Complimentary heat inactivation upon request

Specifications

Additional Assays Biochemical profile, Electrophoretic profile
Certifications/Compliance ISO 13485:2016 certified quality system
Content And Storage 500 mL bottle, store frozen at ≤-10°C
Species Bovine
Age Fetal
Country of Origin Australia
Form Liquid
Format Bottle
Product Type Fetal Bovine Serum
Quantity 500 mL
Serum Treatment Standard (Sterile-filtered)
Sterility Sterile
Sterilization Method Triple-filtered, 0.1 μm
Source Fetal bovine blood
Grade Premium
Processing Triple 0.1 μm sterile-filtered
Tested For Mycoplasma, Sterility (bacteria and fungi), Adventitious viruses
Formulation Liquid, frozen
For Use With (Application) Cell Culture, Biomanufacturing
Heat Inactivated No
Lyophilized No
Product Line GemCell™
Mycoplasma Detected No
Shipping Condition Dry Ice
Regulatory Status RUO
Gamma-irradiated No
Show More Show Less

For cell culture, further manufacturing or research use only. Not for direct therapeutic use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.